3,6-Dichloro-N-methylpyridine-2-carboxamide

Supplier: Apollo Scientific

APOSOR111551-1GEA 0 CHF
APOSOR111551-1G
3,6-Dichloro-N-methylpyridine-2-carboxamide
3,6-Dichloro-N-methylpyridine-2-carboxamide
Formula: C₇H₆Cl₂N₂O
MW: 205.041 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11147606
CAS Number: 1021077-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR