Ethyl 3-amino-4-methoxy-1-benzothiophene-2-carboxylate

Supplier: Apollo Scientific
Warning

APOSOR110275-1GEA 280 CHF
APOSOR110275-1G
Ethyl 3-amino-4-methoxy-1-benzothiophene-2-carboxylate
Ethyl 3-amino-4-methoxy-1-benzothiophene-2-carboxylate
Formula: C₁₂H₁₃NO₃S
MW: 251.302 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13194590
CAS Number: 77373-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR