4-Acetyl-3-(4-chlorophenyl)-5-methylisoxazole

Supplier: Apollo Scientific

APOSOR01994-10GEA 0 CHF
APOSOR01994-10G APOSOR01994-25G
4-Acetyl-3-(4-chlorophenyl)-5-methylisoxazole
4-Acetyl-3-(4-chlorophenyl)-5-methylisoxazole
Formula: C₁₂H₁₀ClNO₂
MW: 235.666 g/mol
Melting Pt: 54…56 °C
Storage Temperature: Ambient
MDL Number: MFCD01313507
CAS Number: 169814-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR