ω-Bromo-2-nitroacetophenone

Supplier: Molekula

Synonyms: Bromomethyl o-nitrophenyl ketoneα-Bromo-2-nitroacetophenone2-Nitrophenacyl bromide2-Bromo-2'-nitroacetophenone

MOLEM19653494EA 0 CHF
MOLEM19653494
ω-Bromo-2-nitroacetophenone
ω-Bromo-2-nitroacetophenone
Formula: C₈H₆BrNO₃
MW: 244.04 g/mol
MDL Number: MFCD00010294
CAS Number: 6851-99-6
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR