Methyl-3-hydroxybenzoate 98%

Supplier: Thermo Scientific
Warning

Synonyms: Methyl 3-Hydroxybenzoate3-Hydroxybenzoic acid methyl ester

ACRO352000250EA 56.9 CHF
ACRO352000250 ACRO352001000
Methyl-3-hydroxybenzoate 98%
Methyl 3-hydroxybenzoate
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 280…281 °C (709 mmHg)
Melting Pt: 70…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00002295
CAS Number: 19438-10-9
EINECS: 243-071-5

Order Now

Specification Test Results

Appearance White to beige Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 70 to 74 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR