Trifluoroacetic acid 0.1% (v/v) in aqueous solution, LiChrosolv®, hypergrade for HPLC, Supelco®

Supplier: Merck

Synonyms: TFA

M-Clarity™ quality level = MQ100

4.80112.9030EA 0 CHF
4.80112.9030 4.80112.2500 4.80112.4000
Trifluoroacetic acid 0.1% (v/v) in aqueous solution, LiChrosolv®, hypergrade for HPLC, Supelco®
Trifluoroacetic acid

LiChrosolv® hypergrade is accurately tested for LC-MS suitability, meeting all the requirements of modern LC-MS ionisation methods (ESI / APCI - positive and negative mode).


Thanks to their low level of ionic background and low ion suppression, LiChrosolv® hypergrade solvents provide high reproducibility and high ionisation efficiency. The packaging material has been improved to perfectly meet LC-MS quality requirements.

Formula: C₂HF₃O₂
MW: 114.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004169
CAS Number: 76-05-1
EINECS: 200-929-3
https://ch.vwr.com/stibo/web/std.lang.all/83/92/728392.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR