Water with 0.1% (v/v) acetic acid, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
M-Clarity™ quality level = MQ100
1.59007.2500EA 77.2 CHF
1.59007.2500
Water with 0.1% (v/v) acetic acid, LiChrosolv®, hypergrade for LC-MS, Supelco®
Water with 0,1% (v/v) acetic acid
LiChrosolv® hypergrade is accurately tested for LC-MS suitability, meeting all the requirements of modern LC-MS ionisation methods (ESI / APCI - positive and negative mode).
Thanks to their low level of ionic background and low ion suppression, LiChrosolv® hypergrade solvents provide high reproducibility and high ionisation efficiency. The packaging material has been improved to perfectly meet LC-MS quality requirements.
Mehr About VWR