Dodecacarbonyltriiron 99% stabilised

Warning

Synonyms: Triiron dodecacarbonylIron dodecacarbonyl

ACRO209290250EA 292 CHF
ACRO209290250 ACRO209290050
Dodecacarbonyltriiron 99% stabilised
Dodecacarbonyltriiron
Formula: C₁₂Fe₃O₁₂
MW: 503.66 g/mol
Melting Pt: 140 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00135617
CAS Number: 17685-52-8
UN: 3178
ADR: 4.1,II

Order Now

Specification Test Results

Appearance Black Powder or crystals
Infrared spectrum Conforms
Stabilizer 1 to 10 % methanol
Titration with Na2S2O3 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR