4-Bromo-3-nitrobenzoic acid 95%

Supplier: Thermo Scientific
Warning

H61975.06EA 126 CHF
H61975.06 H61975.14
4-Bromo-3-nitrobenzoic acid 95%
4-Bromo-3-nitrobenzoic acid
Formula: C₇H₄BrNO₄
MW: 246.02 g/mol
Melting Pt: 199…206 °C
Storage Temperature: Ambient
MDL Number: MFCD00272137
CAS Number: 6319-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR