2-Hydroxy-4-(methacryloyloxy)benzophenone ≥99%

Supplier: Thermo Scientific
Warning

B22445.22EA 2140 CHF
B22445.22 B22445.14 B22445.06
2-Hydroxy-4-(methacryloyloxy)benzophenone ≥99%
2-Hydroxy-4-(methacryloyloxy)benzophenone
Formula: C₁₇H₁₄O₄
MW: 282.30 g/mol
Melting Pt: 77…79 °C
Density: 1.18
MDL Number: MFCD00080561
CAS Number: 2035-72-5
EINECS: 218-000-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR