Potassium chloride 1 M in aqueous solution

Supplier: Thermo Scientific

J63739.AEEA 127 CHF
J63739.AE J63739.AK
Potassium chloride 1 M in aqueous solution
Potassium chloride
Formula: KCl
MW: 74.55 g/mol
Density: 1.13 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR