Capsaicin, Calbiochem®

Supplier: Merck Millipore Calbiochem

Synonyms: 8-Methyl-N-vanillyl-6-none-amid

211275-100MGEA 224 CHF
211275-100MG
Capsaicin, Calbiochem®
Capsaicin
Formula: C₁₈H₂₇NO₃
MW: 305.42 g/mol
Boiling Pt: 210…220 °C (0.01 mmHg)
Melting Pt: 66.1 °C
MDL Number: MFCD00017259
CAS Number: 404-86-4
EINECS: 206-969-8
UN: 2811
ADR: 6.1,III
Merck Index: 12,01811

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR