2-Bromo-5-fluorobenzoic acid

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS20168G100EA 87 CHF
ACTIAS20168G100
2-Bromo-5-fluorobenzoic acid
2-Bromo-5-fluorobenzoic acid
Formula: C₇H₄BrFO₂
MW: 219.01 g/mol
MDL Number: MFCD00142874
CAS Number: 394-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR