3,4,5-Tribromo-1H-pyrazole ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 3,4,5-Tribromopyrazole

A12031.06EA 69.4 CHF
A12031.06 A12031.14
3,4,5-Tribromo-1H-pyrazole ≥97%
3,4,5-Tribromo-1H-pyrazole
Formula: C₃HBr₃N₂
MW: 304.78 g/mol
Melting Pt: 184…188 °C
Storage Temperature: Ambient
MDL Number: MFCD00040248
CAS Number: 17635-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR