Methyl-3-aminosalicylate 97%

Supplier: Thermo Scientific
Warning

Synonyms: Methyl 3-amino-2-hydroxybenzoate

H33035.06EA 732 CHF
H33035.06 H33035.03
Methyl-3-aminosalicylate 97%
Methyl 3-aminosalicylate
Formula: C₈H₉NO₃
MW: 167.17 g/mol
Melting Pt: 79…82 °C
Storage Temperature: Ambient
MDL Number: MFCD09842614
CAS Number: 35748-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR