2-Acetyl-3-amino-5-phenylthiophen 97%

Achtung

B20751.02EA 104 CHF
B20751.02
2-Acetyl-3-amino-5-phenylthiophen 97%
2-Acetyl-3-amino-5-phenylthiophen
Formel: C₁₂H₁₁NOS
MW: 217,29 g/mol
Melting Pt: 119…122 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00068132
CAS-Nummer: 105707-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR