4-Chlor-8-methylchinolin
Lieferant: Thermo Scientific
|
Achtung
|
H50201.03EA
553
CHF
H50201.03
4-Chlor-8-methylchinolin
4-Chloro-8-methylquinoline
Formel:
C₁₀H₈ClN MW: 177,63 g/mol Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00272330 CAS-Nummer: 18436-73-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...