3-Chlorthiophenol
Lieferant: Molekula
Synonyms:
1-Chlor-3-mercaptobenzol
, 3-Chlorphenylmercaptan
, m-Chlorthiophenol
, 3-Chlorbenzolthiol
, 3-Chlorbenzol-1-thiol
MOLE62111567-5GEA
0
CHF
MOLE62111567-5G
3-Chlorthiophenol
3-Chlorthiophenol
Formel:
C₆H₅ClS MW: 144,62 g/mol Siedepunkt: 110 °C (1013 hPa) Dichte: 1,2637 g/cm³ (12.5 °C) |
MDL Number:
MFCD00004839 CAS-Nummer: 2037-31-2 EINECS: 218-010-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...