3-(Trifluormethyl)benzolsulfonamid ≥97%

Lieferant: Thermo Scientific
Gefahr

Synonyms: 3-(Trifluormethyl)benzolsulfonsäureamidα,α,α-Trifluor-m-toluolsulfonamid

L19715.06EA 416 CHF
L19715.06
3-(Trifluormethyl)benzolsulfonamid ≥97%
3-(Trifluormethyl)benzolsulfonamid
Formel: C₇H₆F₃NO₂S
MW: 225,19 g/mol
Melting Pt: 122…126 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00042421
CAS-Nummer: 672-58-2
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR