2-Fluor-5-methylbenzolsulfonylchlorid ≥97%

Gefahr

Synonyms: 2-Fluor-5-methylbenzolsulfonsäurechlorid2-Fluor-5-methylbenzol-1-sulfonylchlorid2-Fluor-5-methylbenzolsulfochlorid

B25264.06EA 195 CHF
B25264.06
2-Fluor-5-methylbenzolsulfonylchlorid ≥97%
2-Fluor-5-methylbenzolsulfonylchlorid
Formel: C₇H₆ClFO₂S
MW: 208,64 g/mol
Melting Pt: 32…34 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD04039785
CAS-Nummer: 870704-14-6
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR