Spirit Thermometer, Clifton
Lieferant: Nickel-Electro
462-0071EA 294 CHF
NICKTC-1
Spirit Thermometer, Clifton
Thermometer clip complete with bent stem spirit filled thermometer Scale: 0 to 100 °C in 2 °C graduations.
  • Sticky pads are fitted to the body of the clip preventing it from scratching the water bath
  • Fitted with two rubber O rings ensuring a snug fit inside the clip, reducing the chances of breakages
  • Temperatures can be easily read even when a SL range accessory lid is fitted
Used as a reference to measure actual water temperature of the water bath.