Benzoxazol
Lieferant: Molekula
MOLE21061250-100GEA
0
CHF
MOLE21061250-100G
Benzoxazol
Benzoxazole
Formel:
C₇H₅NO MW: 119,12 g/mol Siedepunkt: 182…185 °C (1013 hPa) Melting Pt: 28…30 °C Dichte: 1,1754 g/cm³ (20 °C) |
MDL Number:
MFCD00005765 CAS-Nummer: 273-53-0 EINECS: 205-988-9 UN: 1325 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...