5-Methyl-3-isoxazolcarbonsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
L17577.06EA
138
CHF
L17577.06
5-Methyl-3-isoxazolcarbonsäure ≥98%
5-Methylisoxazole-3-carboxylic acid
Formel:
C₅H₅NO₃ MW: 127,1 g/mol Melting Pt: 171…173 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD01318162 CAS-Nummer: 3405-77-4 EINECS: 222-289-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...