Sie suchten nach: Protein- Expression
24  results were found
SearchResultCount:"24"
Sort Results
Listenansicht Easy View
Bewerten Sie das Suchergebnis
Artikel-Nr: (AGLS214303)
Lieferant: AGILENT
Hersteller Artikel Nummer : 214303
Beschreibung: The affinity protein expression and purification system provides a simple, gentle and effective method for high-level protein expression and purificat...
UOM: 1 * 10 mL
Lieferant: Rockland Immunochemicals
Beschreibung: Freund's incomplete adjuvant
Lieferant: AGILENT
Beschreibung: Protein directed evolution in just one step.
Artikel-Nr: (PIER77140)
Lieferant: Thermo Fisher Scientific
Hersteller Artikel Nummer : 77140
Beschreibung: Imject™ Freund's Complete Adjuvant (FCA) is a popular non specific stimulator of the immune response for use in antibody production immunisation proto...
UOM: 1 * 1 SET
Lieferant: Thermo Fisher Scientific
Beschreibung: The Thermo Scientific 1-Step Human High-Yield In Vitro Translation (IVT) Kits enable the expression of functional proteins with 10 to 100 times greate...
Artikel-Nr: (PIER77145)
Lieferant: Thermo Fisher Scientific
Hersteller Artikel Nummer : 77145
Beschreibung: Imject™ Freund's Incomplete Adjuvant (FIA) is a popular non specific stimulator of the immune response for use in antibody production immunisation pro...
UOM: 1 * 1 SET
Lieferant: Thermo Fisher Scientific
Beschreibung: The 1-Step Human Coupled IVT Kit – DNA is a mammalian in vitro translation (IVT) system based on HeLa cell lysates. The kit contains all of the...
Artikel-Nr: (PIER88892)
Lieferant: Thermo Fisher Scientific
Hersteller Artikel Nummer : 88892
Beschreibung: The 1-Step Human High-Yield In Vitro Translation (IVT) Kits enable the expression of functional proteins with 10 to 100 times greater yield per...
UOM: 1 * 1 KIT
Artikel-Nr: (71366-4)
Lieferant: Merck Millipore (Novagen)
Hersteller Artikel Nummer : 71366-4
Beschreibung: The Overnight Express™ Autoinduction Systems enable regulated protein expression in E. coli, without monitoring the culture or adding inducer d...
UOM: 1 * 1 ST
Lieferant: Thermo Fisher Scientific
Beschreibung: The Pierce™ Rapid isotyping kit is a 5-minute lateral-flow assay with ELISA sensitivity for class and subclass determination of monoclonal antibodies.
Artikel-Nr: (71789-3)
Lieferant: Merck Millipore (Novagen)
Hersteller Artikel Nummer : 71789-3
Beschreibung: The Overnight Express™ Autoinduction Systems enable regulated protein expression in E. coli, without monitoring the culture or adding inducer d...
UOM: 1 * 3 SET
Artikel-Nr: (NA9120-1ML)
Lieferant: Cytiva
Hersteller Artikel Nummer : NA9120-1ML
Beschreibung: Protein A conjugated with Horseradish peroxidase for detection of immunoglobulins via affinity to the Fc region - typically IgG from many mammalian sp...
UOM: 1 * 1 mL
Artikel-Nr: (ICNA08642851)
Lieferant: MP Biomedicals
Hersteller Artikel Nummer : 08642851
Beschreibung: Freund's complete adjuvant is made by a modification of Freund's technique, in which 25 mg of Mycobacterium is suspended in a mixture of 7,5 ml...
UOM: 1 * 50 mL
Artikel-Nr: (ICNA08642861)
Lieferant: MP Biomedicals
Hersteller Artikel Nummer : 08642861
Beschreibung: Freund's incomplete adjuvant mixture contains mineral oil and Arlacel A.
UOM: 1 * 50 mL
Lieferant: BIOTECHRABBIT GMBH
Beschreibung: The Rapid Translation System (RTS) is a scalable in vitro protein expression platform that produces large amounts of protein for characterisati...
Lieferant: BIOTECHRABBIT GMBH
Beschreibung: The Rapid Translation System (RTS) is a scalable in vitro protein expression platform that produces large amounts of protein for characterisati...
Inquire for Price
Lager für diesen Artikel ist begrenzt, kann aber in einem Lagerhaus in Ihrer Nähe zur Verfügung. Bitte stellen Sie sicher, dass Sie in sind angemeldet auf dieser Seite, so dass verfügbare Bestand angezeigt werden können. Wenn das call noch angezeigt wird und Sie Hilfe benötigen, rufen Sie uns an 044 745 13 13.
Lager für diesen Artikel ist begrenzt, kann aber in einem Lagerhaus in Ihrer Nähe zur Verfügung. Bitte stellen Sie sicher, dass Sie in sind angemeldet auf dieser Seite, so dass verfügbare Bestand angezeigt werden können. Wenn das call noch angezeigt wird und Sie Hilfe benötigen, rufen Sie uns an 044 745 13 13.
Dieses Produkt kann nur an eine Lieferadresse versandt werden die über die entsprechende Lizenzen verfügt. Für weitere Hilfe bitte kontaktieren Sie Regulatory Affairs.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Das Produkt kann nicht in den Warenkorb gelegt werden.
Der Grund kann ein Preis auf Anfrage oder ein Artikelblock sein.
Bitte kontaktieren Sie Ihr zuständiges VWR Team für weitere Informationen
Dieses Produkt ist Ersatz für den von Ihnen gewünschten Artikel.
Mit diesem Symbol gekennzeichnete Produkte werden nicht mehr angeboten - bis zum Ende des Lagerbestands verkauft. Alternativen können durch Suchen mit der oben aufgeführten VWR-Katalognummer verfügbar sein. Wenn Sie weitere Hilfe benötigen, rufen Sie bitte den VWR-Kundendienst an unter 044 745 13 13
1 - 16 of 24
no targeter for Bottom