Eisen(II)acetat 95%
|
Achtung
|
ACRO305370500EA
378
CHF
ACRO305370500
ACRO305372500
ACRO305370100
Eisen(II)acetat 95%
Eisen(II)acetat
Formel:
C₄H₆FeO₄ MW: 173,94 g/mol Melting Pt: 190 °C (Zersetzung) Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00058909 CAS-Nummer: 3094-87-9 EINECS: 221-441-7 |
Spezifikation Testergebnisse
Appearance | Off-white to light grey to light brown Powder |
Infrared spectrum | Conforms |
Titration with KMnO4 | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...