Biphenyl-4-ol

Lieferant: Molekula

Synonyms: 4-Hydroxybiphenyl4-Phenylphenol4-Hydroxydiphenyl1-Hydroxy-4-phenylbenzol

MOLE35939801-1KGEA 0 CHF
MOLE35939801-1KG
Biphenyl-4-ol
Biphenyl-4-ol
Formel: C₁₂H₁₀O
MW: 170,21 g/mol
Siedepunkt: 321 °C (1013 hPa)
Melting Pt: 162 °C
Dichte: 1,29 g/cm³ (20 °C)
MDL Number: MFCD00002347
CAS-Nummer: 92-69-3
EINECS: 202-179-2
Merck Index: 12,07459

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR