2',4'-Dichlor-5'-fluoracetophenon

Lieferant: Molekula

MOLEM89742438EA 0 CHF
MOLEM89742438
2',4'-Dichlor-5'-fluoracetophenon
2',4'-Dichlor-5'-fluoracetophenon
Formel: C₈H₅Cl₂FO
MW: 207,03 g/mol
MDL Number: MFCD00077499
CAS-Nummer: 704-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR