(R)-(-)-2-Amino-3-methyl-1-butanol

Lieferant: Molekula

Synonyms: D(-)-2-Amino-3-methyl-1-butanolD(-)-Valinol

MOLE37981948-1GEA 0 CHF
MOLE37981948-1G
(R)-(-)-2-Amino-3-methyl-1-butanol
(R)-(-)-2-Amino-3-methyl-1-butanol
Formel: C₅H₁₃NO
MW: 103,16 g/mol
Dichte: 0,93 g/cm³ (20 °C)
MDL Number: MFCD00064297
CAS-Nummer: 4276-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR