Dissolvable Microcarriers, pre-coated
Lieferant: Corning
Corning®
CORN4987EA 166 CHF
CORN4987 CORN4988 CORN4989 CORN4979 CORN4990 CORN4980 CORN4981 CORN4982
Dissolvable Microcarriers, pre-coated
The new Corning Dissolvable Microcarriers are designed to simplify the cell harvest, separation, and concentration processes. All microcarriers arrive pre-sterilized and pre-coated with Corning Synthemax™ II to enhance cell attachment for optimal yield and viability.
  • Dissolvable for easy separation of cells from microcarriers
  • Optical clarity for visualizing cell morphology
  • Sterility Assurance Level (SAL) of 10-6
  • Pre-coated for enhanced cell attachment
  • Nonpyrogenic
They are dissolved within 10 to 20 minutes, allowing for the easy collection of cells without the need for microcarrier separation. Corning Dissolvable Microcarriers offer a cell separation that is faster, gentler, and more convenient than that of standard commercially available microcarriers. Ideal for cell therapy applications, vaccine production and large-volume adherent cell culture scale-up and cell banking.
Bead size: 200 - 300 µm
Surface area: 5000 cm² /g
Lieferumfang: Supplied as dry, sterile powder.