Human DKK3 ELISA Kit

Lieferant: Abnova
ABNOKA1706EA 735 CHF
ABNOKA1706
Human DKK3 ELISA Kit
Human DKK3 ELISA kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human DKK3 in serum, plasma, cell culture supernatants and urine. This assay employs an antibody specific for human DKK-3 coated on a 96-well plate.

  • Detection method: Colorimetric
  • Reactivity: Human
  • Label: Biotin-conjugate

Standards and samples are pipetted into the wells and DKK-3 present in a sample is bound to the wells by the immobilized antibody. The wells are washed and biotinylated anti-human DKK-3 antibody is added. After washing away unbound biotinylated antibody, HRP-conjugated streptavidin is pipetted to the wells. The wells are again washed, a TMB substrate solution is added to the wells and colour develops in proportion to the amount of DKK-3 bound. The stop solution changes the colour from blue to yellow and the intensity of the colour is measured at 450 nm.

Lieferumfang: Kit includes antibody coated microwells (96 wells), 20X wash buffer concentrate (25 ml), standard (2 vials), assay diluent A (30 ml), assay diluent B (15 ml), detection antibody MSP (2 vials), HRP-Streptavidian concentrate (8 µl), TMB one-step substrate reagent (12 ml) and stop solution (8 ml).

Bitte beachten For research use only.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR