Powder dispenser for 24 GC vials
IBSA24-1-250 IBSA24-1-999 IBSA24-1-1000 IBSA24-1-50 IBSA24-1-100
Powder dispenser for 24 GC vials
The 24 GC vial powder dispenser is used to dispense the specific amount of powder in 2 ml autosampler (GC) vials. Can dispense a wide range of solid quantities in a highly precise and repeatable fashion.
  • Simple, fast and reliable
  • High productivity improvement 60-150 x
  • Easy cleaning with 70% ethanol or soap
  • Sterilisable and not autoclavable
  • Ergonomic use as it removes tedious repetitive tasks
  • Custom made volume plates are available
Material: ABS+PC, nylon, RVS304, anodised aluminium
* Customised capacity (µl)
Lieferumfang: Supplied with rack, converter and flight case.