This finest heavy-duty clipper has a powerful universal motor runs at high RPMs and a unique gear-down assembly. This feature gives the clipper greater power for its small size. New larger sized air intake/exhaust openings permit the clipper to run –7 °C cooler than previous clipper models.
- Powerful yet small universal motor
- Up to 2100 SPM provides coarse and fine clipping
- Insulated face plate for cooler running operation
The Cryogen-X™ blades are manufactured from fine grain, high carbon steel. The cutting teeth are machined to exact dimensions for the most precise cutting, and the blades are cryogenically treated for additional wear resistance.
No tools are necessary to change the blades. The clipper has a double insulated twelve foot cord.
Zertifizierungen: UL listed. OSHA and CSA specifications.
Lieferumfang: Supplied with size 40 blade, lubricating oil, grease, cleaning brush, extra set of brush and spring assemblies.