Rust remover, RoVer®
Lieferant: Hach
RoVer®
HACH30001EA 33.1 CHF
HACH30001
Rust remover, RoVer®
Reinigungsmittel Waschmittel und Oberflächenreiniger
This rust remover was developed for cleaning rust-fouled ion-exchange resins. Used to remove rust deposits in lab, shops, laundry and factory applications.