Cold light source, CL 4500 LED CRI90
Lieferant: Zeiss
ZEIS4357009200000EA 0 CHF
ZEIS4357009200000
Cold light source, CL 4500 LED CRI90
The CL 4500 LED CRI90 cold light source is designed for stereomicroscopy.
  • Up to 450 lm light flux at 9 mm fibre output Ø continuous dimming
  • ~5400 K colour temperature and ~90 Color Rendering Index
  • Minimum 50 000 operating hours until intensity drops to 70%
  • 10 mm collet chuck with sensor for 200/1500 light guides
  • Filter slider for 2 filters CL LED and with free opening
  • Two exemplar (stackable), two M4 bushing in bottom
  • Maximum 50 W wide-range supply for flicker-free light, silent axial fan
  • 2,5 mm phone socket for foot button S and country-specific mains cord