CALB324385
324385-1EA 142 CHF
324385-1
CALB324385
A commonly-used broad-spectrum antibiotic. A potent inhibitor of MMP-8 (neutrophil collagenase; IC50 = 30 µM) relative to MMP-1 (interstitial collagenase; IC50 = 300 µM). Also inhibits collagen synthesis by bovine articular chondrocytes cultured in alginate (IC50 = 25 µM). Inhibits nitric oxide synthesis by LPS-activated macrophages without affecting IL-10 release.