Anti-CD48 Mouse Monoclonal Antibody (APC) [clone: 5-4.8]

Lieferant: Biotium
BNCA1057-250EA 426 CHF
BNCA1057-250
Anti-CD48 Mouse Monoclonal Antibody (APC) [clone: 5-4.8]
Reacts with human CD48, a 45 kDa glycosyl phophatidyl-inositol (GPI)-anchored cell surface protein. CD48 is strongly expressed on lymphocytes and monocytes and weakly on granulocytes but is absent on platelets, fibroblasts, epithelium and endothelium. CD48 is one of the markers for detecting the defects of GPI anchoring structure on the patients with paroxysmal nocturnal hemoglobulinuria (PNH) and serves as a low affinity ligand for CD2.

Type: Primary
Antigen: CD48
Clonality: Monoclonal
Clone: 5-4.8
Conjugation: APC
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR