Anti-FCGR3A Mouse Monoclonal Antibody (CF680) [clone: HO-80]

Lieferant: Biotium
BNC801056-500EA 620 CHF
BNC801056-500 BNC801056-100
Anti-FCGR3A Mouse Monoclonal Antibody (CF680) [clone: HO-80]
This antibody recognizes CD16 (FcγRIII), the low-affinity receptor for IgG with an apparent molecular weight of 50-80 kDa. Two similar genes represent CD16, CD16A (FcγRIIIA), which exists as a hetero-oligomeric polypeptide-anchored form in macrophages and NK cells and CD16B (FcγRIIIB), which exist as a monomeric GPI-anchored form in neutrophils. Furthermore, there are two known polymorphisms of CD16B, NA-1 and NA-2. Individuals homozygous for NA-2 show a lower phagocytic capacity compared with NA-1. CD16 binds IgG in the form of immune complexes and shows preferential binding of IgG1 and IgG3 isotypes and minimal binding of IgG2 and IgG4. Upon IgG binding, both CD16 isoforms initiate signal transduction cascades that lead to a variety of responses including antibody-dependent cell-mediated cytotoxicity (ADCC), phagocytosis, degranulation and proliferation.

Type: Primary
Antigen: FCGR3A
Clonality: Monoclonal
Clone: HO-80
Conjugation: CF680
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR