Anti-HLA-A; HLA-B; HLA-C Mouse Monoclonal Antibody (CF543) [clone: 246-B8.E7]

Lieferant: Biotium
BNC431054-500EA 620 CHF
BNC431054-500 BNC431054-100
Anti-HLA-A; HLA-B; HLA-C Mouse Monoclonal Antibody (CF543) [clone: 246-B8.E7]
Reacts with a monomorphic determinant of human major histocompatibility (MHC) class I antigens (HLA-A, B and C). Human MHC class I antigens are expressed constitutively on all nucleated cells lymphocytes such as lymphocytes, thymocytes, granulocytes, and bone marrow cells and are absent on erythrocytes. MHC class I antigens play a role in class I MHC-associated antigen presentation, inhibition of NK cell cytotoxicity, tumor surveillance, and tissue allotransplantation.

Type: Primary
Antigen: HLA-A; HLA-B; HLA-C
Clonality: Monoclonal
Clone: 246-B8.E7
Conjugation: CF543
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human, Mouse, Non-human primates
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR