ICNA02159810
ICNA0215981010EA 242 CHF
ICNA0215981010 ICNA0215981005
ICNA02159810
Rolipram is a selective, cell permeable inhibitor of cAMP-specific phosphodiesterase (PDE4, IC50=1 μM).
Rolipram compound has been shown to inhibit PDE4 activity in human cavernosal tissue, which interrupts the signal transduction pathway of erectile response. Rolipram promotes apoptosis in HL60 cells through a cAMP-independent mechanism and has been shown to enhance neuronal survival. It is a cAMP-specific phosphodiesterase inhibitor, selective for phosphodiesterase IV.
Soluble in DMSO, ethanol and acetone (25 mg/ml each).