D-Proline methyl ester hydrochloride 98%

Lieferant: Thermo Scientific
Achtung

ACRO445590050EA 211 CHF
ACRO445590050
D-Proline methyl ester hydrochloride 98%
D-Proline methyl ester hydrochloride
Formel: C₆H₁₁NO₂
MW: 165,62 g/mol
Melting Pt: 42…49 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD00083685
CAS-Nummer: 65365-28-8

Order Now

Spezifikation Testergebnisse

Appearance White Powder or solid
Infrared spectrum Conforms
Titration Argentometric ≥97.5 to ≤102.5 %
Specific optical rotation +31° to +35° (20°C, 589 nm) (c=1 in H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR