(+)-Griseofulvin 97%

Gefahr

ACRO227450050EA 29.7 CHF
ACRO227450050
(+)-Griseofulvin 97%
(+)-Griseofulvin

Appearance: White to yellowish-white Powder

Formel: C₁₇H₁₇ClO₆
MW: 352,77 g/mol
Melting Pt: 217…224 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD00082343
CAS-Nummer: 126-07-8

Order Now

Spezifikation Testergebnisse

Appearance White to yellowish-white Powder
Infrared spectrum Conforms
Melting point 217 to 224 °C
Loss on drying ≤1 % (1 g, 105 °C)
UV spectrophotometry ≥96.0 % (on dried substance)
Specific optical rotation +354° to +364° (20 °C, 589 nm) (c=1, DMF) (on dried substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR