D-erythro-Sphingosine ≥99%

Lieferant: Thermo Scientific

J63584.MBEA 422 CHF
J63584.MB
D-erythro-Sphingosine ≥99%
D-erythro-Sphingosine

D-erythro-Sphingosine is used to inhibit protein kinase C and calmodulin-dependent enzymes, induces apoptosis. And also used to constituent of cell membranes.


Bitte beachten Store at –20 °C.

Formel: C₁₈H₃₇NO₂
MW: 299,50 g/mol
MDL Number: MFCD00036751
CAS-Nummer: 123-78-4
EINECS: 204-651-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR