Stabilizer for oil based standard solutions

Lieferant: Thermo Fisher Scientific
43871.18EA 254 CHF
ALFA43871.18
Stabilizer for oil based standard solutions
This stabilizer added to solutions at 0,6 wt%.

  • Physical form: liquid
  • MDL No.: MFCD00148498
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR