Anti-AAMP Rabbit Polyclonal Antibody

Supplier: Bioss
BOSSBS-11388REA 920 CHF
BOSSBS-11388R
Anti-AAMP Rabbit Polyclonal Antibody
Antibodies
AAMP is a 434 amino acid immunoglobulin-like protein that contains 8 WD repeats. Expressed in endothelial cells, cytotrophoblasts and blood vessels, AAMP is thought to have a heparin-sensitive role in cell adhesion and cell migration. AAMP is strongly expressed in poorly differentiated colon adenocarcinoma cells, suggesting a role for AAMP in tumor progression.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: AAMP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR