Anti-TPOR Rabbit Polyclonal Antibody

Supplier: Bioss
BOSSBS-11311REA 920 CHF
BOSSBS-11311R
Anti-TPOR Rabbit Polyclonal Antibody
Antibodies
Thrombopoietin (TPO or THPO), also known as c-Mpl ligand (c-Mpl L), is a cytokine that plays a central role in megakaryopoiesis by influencing the development and maturation of megakaryocytes and platelet production from hematopoietic stem cells. TPO exerts its biological effects through the TPO receptor, c-Mpl. c-Mpl is a member of the cytokine receptor superfamily. Expression of c-Mpl is restricted to hematopoietic tissues and cells, such as bone marrow, spleen, fetal liver and CD34+ cells. Stimulation of c-Mpl with TPO results in the activation of the Janus tyrosine kinase family members, Tyk 2 and JAK2, which in turn phosphorylate Stat5 and Stat3, causing their nuclear translocation and the transcription of Stat responsive genes. Muta-tions in c-Mpl have been implicated as the cause of certain human disorders, including congenital amegakaryocytic thrombocytopenia (CAMT) and thrombocytopenia with absent radii (TAR) syndrome.

WB(1:100-1000), FCM(1:20-100), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: TPOR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR