Anti-SRD5A1 Rabbit Polyclonal Antibody

Lieferant: Bioss
BOSSBS-11308REA 920 CHF
BOSSBS-11308R
Anti-SRD5A1 Rabbit Polyclonal Antibody
Antikörper
Steroid 5a-Reductase is an important enzyme in androgen physiology because it catalyzes the conversion of testosterone into the more potent 5a-dihydro-testosterone, which mediates androgen effects on target tissues. The enzyme exists as two isoforms: type 1, which is expressed mainly in the skin; and type 2, which is expressed mainly in the prostate. In cultured human skin cells, 5a-Reductase 1 shows heterogeneity of protein, and has different levels of transcriptional and translational expression. 5a-Reductase 1 is expressed in all portions of the hair follicle, whereas 5a-Reductase 2 is expressed only in mesenchymal portions. In addition, 5a-Reductase 1 is mainly expressed in human breast carcinoma and may play a role in the in situ production and actions of the potent androgen 5a-dihydrotestosterone, including inhibition of cancer cell proliferation in hormone-dependent human breast carcinoma. The 5a-Reductase-3a-hydroxysteroid dehydrogenase complex is present in the human brain, suggesting that the complex may be involved in the synthesis of neuroactive steroids or the catabolism of neurotoxic steroids.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: SRD5A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR