Anti-SPRR3 Rabbit Polyclonal Antibody

Supplier: Bioss
BOSSBS-11163REA 920 CHF
BOSSBS-11163R
Anti-SPRR3 Rabbit Polyclonal Antibody
Antibodies
Esophagin, also known as small proline-rich protein 3 (SPR3) or Cornifin ∫, belongs to the cornifin family of cornified-envelope structural proteins. It is expressed in mucosal epithelia such as esophagus and tongue and is strongly induced during epidermal keratinocyte differentiation. Due to its highly inducible nature, Esophagin is considered a marker of squamous differentiation. Esophagin serves as a cross-linking protein within the cornified cell envelope and may play a role in the maintenance of normal esophageal epithelial homeostasis. It shares significant homology with the related proteins, SPRR1 and SPRR2. Esophagin is typically not expressed in healthy human epithelium, but its expression is upregulated in numerous hyperproliferative disorders of the skin. Contrastly, its expression is dramatically downregulated in esophageal squamous cell carcinoma.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: SPRR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR