Anti-RRAGC Rabbit Polyclonal Antibody

Supplier: Bioss
BOSSBS-11111REA 920 CHF
BOSSBS-11111R
Anti-RRAGC Rabbit Polyclonal Antibody
Antibodies
Protocadherins are a subfamily of cadherins, a large group of related glycoproteins that mediate calcium-dependent cell-to-cell adhesion via a homophilic mechanism. Involved in a variety of functions, protocadherins help to regulate neural development and synapse formation. PCDH1 is a 1,026 amino acid single-pass type I membrane protein that contains seven cadherin domains and is a member of the protocadherin family. Localized to cell-cell and cell-matrix boundaries and expressed at high levels in brain and neuro-glial cells, PCDH1 is thought to be involved in cell adhesion and cell-cell interactions and may play a role in neuronal development. PCDH1 contains a C-terminal cytoplasmic region, an extracellular region and a transmembrane region, and is expressed as two isoforms due to alternative splicing events.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: RRAGC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR