Anti-VARS2 Rabbit Polyclonal Antibody

Supplier: Bioss
BOSSBS-10504REA 920 CHF
BOSSBS-10504R
Anti-VARS2 Rabbit Polyclonal Antibody
Antibodies
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: VARS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR