Anti-SYNGAP Rabbit Polyclonal Antibody

Supplier: Bioss
BOSSBS-10392REA 614 CHF
BOSSBS-10392R
Anti-SYNGAP Rabbit Polyclonal Antibody
Antibodies
The PSD-95/SAP 90 family of proteins, which are known to bind to and cluster various membrane proteins, are involved in the organization of synaptic structure. SynGAP (for synaptic GTPase activating protein), a novel synaptic RasGAP, is a brain-specific protein abundant in the cortex, hippocampus and olfactory bulb. SynGAP interacts with all three PDZ domains within PSD-95/ SAP 90 proteins. It represents one of three classes of mammalian RasGAPs and is specifically localized to excitatory synapses possessing NMDA receptors. SynGAP may be involved in the regulation of BDNF as well as Ras signaling. Its activity is inhibited by phosphorylation by CaMKII, which could result in the activation of the MAP kinase pathway.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: SYNGAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR